Learn More
Description
Specifications
Specifications
| Assay Range | 4.69 to 300 pg/mL |
| Assay Sensitivity | 0.696 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
| Specificity | Natural and Recombinant Mouse ELA2 |
| Gene ID (Entrez) | 2668 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50 μL),Cell Lysates (50 μL),Tissue Lysates (50 μL),Serum (50 μL),EDTA Plasma (50 μL),Heparin Plasma (50 μL) |
| Storage Requirements | Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Total Assay Time | 4 hr. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
