Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GDNF Recombinant Protein

Catalog No. PIRP8602
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8602 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8602 Supplier Invitrogen™ Supplier No. RP8602

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Recombinant Protein

GDNF (human) Recombinant Protein, RP-8602, from Invitrogen.

Glial Cell Line-Derived Neurotrophic Factor (GDNF) is a neurotrophic factor that is closely related to other neurotrophic factors, such as Neurturin, Persephin, and Artemin, by a common structural feature called the cysteine-knot. GDNF signals through a multicomponent system of receptors that includes RET and GFRalpha1-4, to promote dopamine uptake, survival and differentiation of neurons. Recombinant human GDNF is a non-glycosylated homodimer, containing two 135 amino acid chains, with a total molecular weight of 30.4 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of C6 cells and is typically 0.5-5 ug/mL. The specific activity of this protein is 2x10^3 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39905
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.01M citric acid with 0.1M NaCl and no preservative; pH 4.0
Gene ID (Entrez) 2668
Molecular Weight (g/mol) 30.36
Name Human Glial Cell Derived Neurotrophic Factor (GDNF)
pH Range 4.0
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-GDNF
Common Name Human Glial Cell Derived Neurotrophic Factor (GDNF)
Gene Symbol GDNF
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research purpose only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.