Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GDPD1 Full-length ORF (AAH34432.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-751 25 μg
89-945-750 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-751 Supplier Abnova™ Supplier No. H00284161P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the glycerophosphodiester phosphodiesterase family of enzymes that catalyze the hydrolysis of deacylated glycerophospholipids to glycerol phosphate and alcohol. The encoded protein is localized to the cytoplasm and concentrates near the perinuclear region. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MSSTAAFYLLSTLGGYLVTSFLLLKYPTLLHQRKKQRFLSKHISHRGGAGENLENTMAAFQHAVKIGTDMLELDCHITKDEQVVVSHDENLKRATGVNVNISDLKYCELPPYLGKLDVSFQRACQCEGKDNRIPLLKEVFEAFPNTPINIDIKVNNNVLIKKVSELVKRYNREHLTVWGNANYEIVEKCYKENSDIPILFSLQRVLLILGLFFTGLLPFVPIREQFFEIPMPSIILKLKEPHTMSRSQKFLIWLSDLLLMRKALFDHLTARGIQVSFWNDAFWKQHSSPV

Specifications

Accession Number AAH34432.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 284161
Molecular Weight (g/mol) 59.8kDa
Name GDPD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSSTAAFYLLSTLGGYLVTSFLLLKYPTLLHQRKKQRFLSKHISHRGGAGENLENTMAAFQHAVKIGTDMLELDCHITKDEQVVVSHDENLKRATGVNVNISDLKYCELPPYLGKLDVSFQRACQCEGKDNRIPLLKEVFEAFPNTPINIDIKVNNNVLIKKVSELVKRYNREHLTVWGNANYEIVEKCYKENSDIPILFSLQRVLLILGLFFTGLLPFVPIREQFFEIPMPSIILKLKEPHTMSRSQKFLIWLSDLLLMRKALFDHLTARGIQVSFWNDAFWKQHSSPV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ27503/FLJ37451/GDE4/MGC35046/UGPQ
Common Name GDPD1
Gene Symbol GDPD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.