Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GFI1 Partial ORF (NP_005254, 1 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945774 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-774 10 ug
89-945-775 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-774 Supplier Abnova™ Supplier No. H00002672Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a nuclear zinc finger protein that functions as a transcriptional repressor. This protein plays a role in diverse developmental contexts, including hematopoiesis and oncogenesis. It functions as part of a complex along with other cofactors to control histone modifications that lead to silencing of the target gene promoters. Mutations in this gene cause autosomal dominant severe congenital neutropenia, and also dominant nonimmune chronic idiopathic neutropenia of adults, which are heterogeneous hematopoietic disorders that cause predispositions to leukemias and infections. Multiple alternatively spliced variants, encoding the same protein, have been identified for this gene. [provided by RefSeq]

Sequence: MPRSFLVKSKKAHSYHQPRSPGPDYSLRLENVPAPSRADSTSNAGGAKAEPRDRLSPESQLTEAPDRASASPRQLRSSVCERSSEFEDFWR

Specifications

Accession Number NP_005254
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2672
Molecular Weight (g/mol) 35.75kDa
Name GFI1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MPRSFLVKSKKAHSYHQPRSPGPDYSLRLENVPAPSRADSTSNAGGAKAEPRDRLSPESQLTEAPDRASASPRQLRSSVCERSSEFEDFWR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ94509/GFI-1/ZNF163
Common Name GFI1
Gene Symbol GFI1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.