Learn More
Invitrogen™ Human GFR alpha-1/GDNF R ELISA Kit
Description
Human GFR alpha-1/GDNF R quantitates human GFR alpha-1/GDNF R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GFR alpha-1/GDNF R.
Specifications
Specifications
| Accession Number | AU042498; DKFZp313E1012; DKFZp686J0156; FLJ10561; FLJ31546; GDNF family receptor alpha 1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR; GDNFRA; GDNFR-alpha; GDNFR-alpha-1; gfr alpha 1; GFR alpha-1; GFRA1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; Trnr1 |
| Assay Range | 0.35 to 80 ng/mL |
| Assay Sensitivity | 0.35 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2674 |
| Gene Symbol | GFRA1 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.