Learn More
BioVendor, LLC Human Growth Hormone Binding Protein (GHBP) ELISA, 96 wells
SDP
Supplier:  BioVendor, LLC RMEE024R
Encompass
Human Growth Hormone Binding Protein (GHBP) ELISA, 96 wells. Immunoassay kit to measure GHBP in human serum or plasma samples. Two Control Sera are provided for QC purposes. Cal. range: 0.006-84 ng/mL. Detection limit: 0.006 ng/mL. SUMMARY: In humans, GHBP is formed by receptor shedding of GH receptor (Somatotropin receptor) by ADAM17. In equilibrium ~50% of circulating GH is bound to GHBP. In guinea pig model, complex formation increases half-life from 11-20 mins up to ~100 mins and in general binding to GHBP inhibits GH cellular action. GHBP levels are independent of GH pulsatility and do not show circadian rhythm. GHBP correlates positively with intraabdominal fat mass and is increased in T2D with hyperinsulinemia. Undetectable GHBP levels could point to GH insensitivity, caused by a deletion in GHR gene. Further, IGF-I/GHBP ratio may indicate GH-deficiency in adults and may be predictive for GH treatment response. GHBP may be a biomarker for the amount of visceral adipose tissue.
Catalog No. 50-206-9270
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.