Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GHR ELISA Kit

Catalog No. EH219RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH219RB 96 Tests
EH219RBX5 5 x 96 Tests
EH219RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH219RB Supplier Invitrogen™ Supplier No. EH219RB
Add to Cart
Add to Cart

ELISA

Human GHR quantitates human GHR in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GHR.

may play a role in chondrocyte differentiation and positive regulation of longitudinal bond growth and.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number GH receptor; GH-binding protein; GHBP; GHIP; Ghr; GHR/BP; growth hormone binding protein; growth hormone receptor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; MGC124963; MGC156665; serum binding protein; Serum-binding protein; Somatotropin receptor; unnamed protein product
Assay Range 0.4 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2690
Gene Symbol GHR
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Growth Hormone Receptor
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.