Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GIYD1 Full-length ORF (AAI48778.1, 1 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945906 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-906 10 μg
89-945-907 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-906 Supplier Abnova™ Supplier No. H00548593P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

A segmental duplication of the p arm of chromosome 16 created two identical copies of the GIY-YIG domain containing gene, this record represents the more centromeric copy. Exons of this gene overlap with exons of the phenol-preferring sulfotransferase (SULT1A3) gene. Two transcript variants that encode different protein isoforms have been identified through sequence analysis. [provided by RefSeq]

Sequence: MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLET

Specifications

Accession Number AAI48778.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 548593
Molecular Weight (g/mol) 44.66kDa
Name GIYD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MGPAGVAARPGRFFGVYLLYCLNPRYRGRVYVGFTVNTARRVQQHNGGRKKGGAWRTSGRGPWEMVLVVHGFPSSVAALRDEEGPLCCPHPGCLLRAHVICLAEEFLQEEPGQLLPLEGQCPCCEKSLLWGDLIWLCQMDTEKEVEDSELEEAHWTDLLET
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GIYD1
Gene Symbol GIYD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.