Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GJA4 Full-length ORF (NP_002051.2, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89945911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-945-911 25 ug
89-945-910 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-945-911 Supplier Abnova™ Supplier No. H00002701P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene have been associated with atherosclerosis and a higher risk of myocardial infarction. [provided by RefSeq]

Sequence: MGDWGFLEKLLDQVQEHSTVVGKIWLTVLFIFRILILGLAGESVWGDEQSDFECNTAQPGCTNVCYDQAFPISHIRYWVLQFLFVSTPTLVYLGHVIYLSRREERLRQKEGELRALPAKDPQVERALAAVERQMAKISVAEDGRLRIRGALMGTYVASVLCKSVLEAGFLYGQWRLYGWTMEPVFVCQRAPCPYLVDCFVSRPTEKTIFIIFMLVVGLISLVLNLLELVHLLCRCLSRGMRARQGQDAPPTQGTSSDPYTDQVFFYLPVGQGPSSPPCPTYNGLSSSEQNWANLTTEERLASSRPPLFLDPPPQNGQKPPSRPSSSASKKQYV

Specifications

Accession Number NP_002051.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2701
Molecular Weight (g/mol) 63.8kDa
Name GJA4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MGDWGFLEKLLDQVQEHSTVVGKIWLTVLFIFRILILGLAGESVWGDEQSDFECNTAQPGCTNVCYDQAFPISHIRYWVLQFLFVSTPTLVYLGHVIYLSRREERLRQKEGELRALPAKDPQVERALAAVERQMAKISVAEDGRLRIRGALMGTYVASVLCKSVLEAGFLYGQWRLYGWTMEPVFVCQRAPCPYLVDCFVSRPTEKTIFIIFMLVVGLISLVLNLLELVHLLCRCLSRGMRARQGQDAPPTQGTSSDPYTDQVFFYLPVGQGPSSPPCPTYNGLSSSEQNWANLTTEERLASSRPPLFLDPPPQNGQKPPSRPSSSASKKQYV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CX37
Common Name GJA4
Gene Symbol GJA4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.