Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GLI-3 ELISA Kit

Catalog No. EH220RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH220RB 96 Tests
EH220RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH220RB Supplier Invitrogen™ Supplier No. EH220RB
Add to Cart
Add to Cart

ELISA

Human GLI-3 quantitates human GLI-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GLI-3.

This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV
Assay Range 40 to 10,000 pg/mL
Assay Sensitivity 40 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2737
Gene Symbol GLI3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GLI3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.