Learn More
Cell Systems Corporation 5 Vials Primary Human Glomerular Microvascular Endothelial Cells - Same Lot (ACBRI 128V5)
SDP
Supplier:  Cell Systems Corporation ACBRI128V5
Encompass_Preferred
Boost your research program with 5 identical vials of passage 3 Primary Human Glomerular Microvascular Endothelial Cells while saving ~ 20% on purchase. Isolated from normal human kidney cortical tissue without positive control antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.
Catalog No. 50-206-8276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.