Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Glucosylceramidase/GBA Recombinant Protein, Carrier-Free

Catalog No. 7410GH01M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-10G-H01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 74-10G-H01M Supplier R&D Systems™ Supplier No. 7410GH01M
This item is not returnable. View return policy

Defects in Glucosylceramidase are the cause of Gaucher disease, also known as glucocerebrosidase deficiency.

Glucosylceramidase is lysosomal enzyme that cleaves the beta-glucosidic linkage of glucosylceramide, an intermediate in glycolipid metabolism. The mature enzyme has 497 amino acids with a molecular weight of 62 kDa. Glycosylation occurs at four of five N-glycosylation sites and is essential for the trafficking and activity of the enzyme.

Storage:

Use manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, −70°C as supplied. 3 months, −70°C under sterile conditions after opening.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P04062
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT
Gene ID (Entrez) 2629
Molecular Weight (g/mol) 56 kDa
Quantity 1 mg
Storage Requirements Use manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, −70°C as supplied. 3 months, −70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method
Biological Activity >180pmol/min/μg
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.