Learn More
R&D Systems™ Human GluR6/GRIK2 (NP_068775) VersaClone cDNA
SDP
Catalog No. RDC0968 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC0968 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC0968 Supplier R&D Systems™ Supplier No. RDC0968
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glutamate receptor, ionotropic, kainate 2
The Human GluR6/GRIK2 (NP_068775) VersaClone cDNA clone contains the complete ORF for KLHL24, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The KLHL24 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Gene Alias DRE1, EAA4, GLR6, GluK2, GLUK6, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24 KRIP6, MRT6, Protein DRE1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans