Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Glut4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8654SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8654SP 25 μg
MAB8654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8654SP Supplier R&D Systems Supplier No. MAB8654SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glut4 Monoclonal specifically detects Glut4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Glut4
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 925951
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P14672
Gene Alias GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, SLC2A4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols SLC2A4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glut4 Met1-Asp509 Accession # P14672
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6517
Test Specificity Detects human Glut4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.