Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Glutamate Dehydrogenase ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP267990 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267990 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267990 Supplier Novus Biologicals™ Supplier No. NBP267990

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Kit (Chemiluminescence)

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 mIU/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Human GDH/GLDH in samples. No significant cross-reactivity or interference between Human GDH/GLDH and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 2746
Gene Symbol GLUD1
Quantity 1 Kit
Research Discipline Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 1.4.1, EC 1.4.1.3, GDH, GDH 1, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, glutamate dehydrogenase 1, mitochondrial, MGC132003
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.