Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human Glutamate Receptor 1 Peptide, Synthetic, >90% Purity, 1 mg

Catalog No. 502703984
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-3984 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-3984 Supplier Abcam Supplier No. AB284241MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide for use in blocking experiments with >90% purity.

  • Species Reactivity: Human
  • Tested Applications: BL
  • Form: Liquid
  • Purity: >90%
  • Storage Conditions: Store at -20°C
  • Buffer: pH 6.75, containing 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, and 0.001% Sorbitan monolaurate, ethoxylated

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Glutamate Receptor 1 (AMPA subtype), Human
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Reactivity Human
Label or Dye Unconjugated
Form Liquid
Buffer 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75
Content And Storage Store at -20°C.
Product Type Blocker and Diluent
Concentration 1 mg/mL
Cross Reactivity Human
Formulation pH 6.75, 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated
Quantity 1 mg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.