Learn More
Novus Biologicals™ Human Glutathione S-Transferase pi 1/GSTP1 ELISA Kit (Chemiluminescence)

Specifications
Specifications
| Assay Range | 0.47 to 30 ng/mL |
| Assay Sensitivity | 0.28 ng/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Human Glutathione S-Transferase pi 1/GSTP1 in samples. No significant cross-reactivity or interference between Human Glutathione S-Transferase pi 1/GSTP1 and analogues was observed. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Gene ID (Entrez) | 2950 |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.