Learn More
ACROBiosystems Human Glycoprotein 1mg
Supplier:  ACROBiosystems GLNM6451MG
Encompass_Preferred
Rabies virus (RABV) scientific name Rabies lyssavirus is a deadly neurotropic virus that causes rabies in humans and animals. Rabies virus has an extremely wide host range and its transmission most often occur through the saliva of animals. Without intervention prior to disease progression rabies has the highest case fatality of any infectious disease. RABV contains a single-stranded negative-sense RNA genome that encodes five structural proteins: nucleoprotein (N) phosphoprotein (P) matrix protein (M) glycoprotein (G) and RNA-dependent RNA polymerase (L). Among these viral proteins the RABV glycoprotein (RABV-G) is a pivotal player mediating virus entry and the major target of neutralizing antibodies thus a key factor for vaccine and drug design.
Catalog No. 50-253-5006
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.