Learn More
ACROBiosystems Human Glypican 3 (GPC3) R355A,R358A Protein, Fc Tag, HEK293, 100 μg
Supplier: ACROBiosystems GP3H5255100UG
Recombinant human Glypican-3 (GPC3) R355A,R358A is an Fc-tagged, glycosylated protein expressed in HEK293 cells, provided purified for use in binding studies, assay development, and biochemical characterization.
- Mutations: R355A and R358A.
- Tag: Human IgG1 Fc at the C-terminus.
- Expression system: HEK293 (human 293 cells).
- Amino acid range: Gln 25 - His 559.
- Calculated molecular weight: 87.1 kDa; glycosylated protein migrates ~230 kDa on non-reducing SDS-PAGE and is ~170-210 kDa by SEC-MALS.
- Purity: >95% by SDS-PAGE; >90% by SEC-MALS.
- Available sizes: 100 μg and 1 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.