Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
Human GM-CSF Capture Antibody; Human GM-CSF Detection Antibody; Human GM-CSF Standard; Streptavidin-HRP
The Human GM-CSF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human GM-CSF.
Specifications
Specifications
| Conjugate | Biotin |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 5 Plates |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
