Learn More
R&D Systems™ Human GM-CSF Quantikine ELISA Kit

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 3pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | Natural and Recombinant Human GM-CSF |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1437 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.