Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human GM-CSF Recombinant Protein, PeproTech®

Catalog No. 30003500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30003500UG 500 μg
30003100UG 100 μg
300-03-1MG 1 mg
300-03-20UG 20 μg
30003250UG 250 μg
300-03-50UG 50 μg
300-03-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 30003500UG Supplier Gibco™ Supplier No. 30003500UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids, containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04141
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1437
Molecular Weight (g/mol) 14.6 kDa
Name Human GM-CSF
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Common Name GM-CSF
Gene Symbol CSF2
Biological Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.