Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GM130 Synthetic Peptide

Catalog No. PEP296
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP296 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP296 Supplier Invitrogen™ Supplier No. PEP296
Add to Cart
Add to Cart

Synthetic Peptide

GM130/GOLGA2 Synthetic Peptide, PEP-296, from Invitrogen™.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2801
Species Human
Purification Method purified
Accession Number Q08379
Common Name Human GM130
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Lyophilized
Formulation water with no preservative
For Use With (Application) Blocking Assay, Control
Gene Alias Golgin subfamily A member 2, 130 kDa cis-Golgi matrix protein, GM130, GM130 autoantigen, Golgin-95, GOLGA2
Gene Symbol GOLGA2
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.