Learn More
Sino Biological Human GMEB1 PAb 100UL 108011-T10-100
Supplier:  Sino Biological 108011T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GMEB1, and purified by antigen affinity chromatography.
Catalog No. 50-241-2057
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.