Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human GMF-beta Recombinant Protein, PeproTech®

Catalog No. 45037500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45037500UG 500 μg
45037100UG 100 μg
450-37-10UG 10 μg
450-37-1MG 1 mg
45037250UG 250 μg
450-37-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45037500UG Supplier Gibco™ Supplier No. 45037500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GMF-beta is a 16.5 kDa globular protein containing 141 amino acid residues, including one intramolecular disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60983
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2764
Molecular Weight (g/mol) 16.5 kDa
Name Human GMF-beta
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 3110001H22Rik; 3110001O16Rik; AI851627; C79176; D14Ertd630e; DNA for thyroid hormone receptor binding site (276 bp); glia maturation factor beta; glia maturation factor beta-like protein; glia maturation factor, beta; GMF; Gmfb; GMF-beta
Common Name GMFB
Gene Symbol Gmfb
Conjugate Unconjugated
Recombinant Recombinant
Sequence SESLVVCDVA EDLVEKLRKF RFRKETNNAA IIMKIDKDKR LVVLDEELEG ISPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.