Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human GnRH Peptide, Synthetic, >95% Purity, 50 μg

Catalog No. 502710940
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0940 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0940 Supplier Abcam Supplier No. AB4176950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic human GnRH blocking peptide with >95% purity, suitable for neutralization and blocking applications.

  • Peptide Type: Synthetic
  • Purity: >95%
  • Applications: BL, Neut
  • Species Reactivity: Human
  • Form: Lyophilized

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Concentration 0.5 mg/mL
Content And Storage Store at -20°C.
Label or Dye Unconjugated
Format Vial
Form Lyophilized
Target Molecule GnRH (Human)
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Cross Reactivity Human
Quantity 50 μg
For Use With (Application) Blocking, Neutralization
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.