Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GOLGA1 Full-length ORF (AAH32853.1, 1 a.a. - 767 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-215 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-215 Supplier Abnova™ Supplier No. H00002800P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome. [provided by RefSeq]

Sequence: MFAKLKKKIAEETAVAQRPGGATRIPRSVSKESVASMGADSGDDFASDGSSSREDLSSQLLRRNEQIRKLEARLSDYAEQVRNLQKIKEKLEIALEKHQDSSMRKFQEQNETFQANRAKMAEGLALALARKDQEWSEKMDQLEKEKNILTAQLQEMKNQSMNLFQRRDEMDELEGFQQQELSKIKHMLLKKEESLGKMEQELEARTRELSRTQEELMNSNQMSSDLSQKLEELQRHYSTLEEQRDHVIASKTGAESKITALEQKEQELQALIQQLSIDLQKVTAETQEKEDVITHLQEKVASLEKRLEQNLSGEEHVQELLKEKTLAEQNLEDTRQQLLAARSSQAKAINTLETRVRELEQTLQASEEQLQQSKGIVAAQETQIQELAAANQESSHVQQQALALEQQFLERTQALEAQIVALERMRAADQTTAEQGMRQLEQENAALKECRNEYERSLQNHQFELKKLKEEWSQREIVSVAMAQALEEVRKQREEFQQQAANLTAIIDEKEQNLREKTEVLLQKEQEILQLERGHNSALLQIHQLQAELEALRTLKAEEAAVVAEQEDLLRLRGPLQAEALSVNESHVTSRAMQDPVFQLPTAGRTPNGEVGAMDLTQLQKEKQDLEQQLLEKNKTIKQMQQRMLELRKTLQKELKIRPDNELFEVREKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWS

Specifications

Accession Number AAH32853.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2800
Molecular Weight (g/mol) 114.6kDa
Name GOLGA1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MFAKLKKKIAEETAVAQRPGGATRIPRSVSKESVASMGADSGDDFASDGSSSREDLSSQLLRRNEQIRKLEARLSDYAEQVRNLQKIKEKLEIALEKHQDSSMRKFQEQNETFQANRAKMAEGLALALARKDQEWSEKMDQLEKEKNILTAQLQEMKNQSMNLFQRRDEMDELEGFQQQELSKIKHMLLKKEESLGKMEQELEARTRELSRTQEELMNSNQMSSDLSQKLEELQRHYSTLEEQRDHVIASKTGAESKITALEQKEQELQALIQQLSIDLQKVTAETQEKEDVITHLQEKVASLEKRLEQNLSGEEHVQELLKEKTLAEQNLEDTRQQLLAARSSQAKAINTLETRVRELEQTLQASEEQLQQSKGIVAAQETQIQELAAANQESSHVQQQALALEQQFLERTQALEAQIVALERMRAADQTTAEQGMRQLEQENAALKECRNEYERSLQNHQFELKKLKEEWSQREIVSVAMAQALEEVRKQREEFQQQAANLTAIIDEKEQNLREKTEVLLQKEQEILQLERGHNSALLQIHQLQAELEALRTLKAEEAAVVAEQEDLLRLRGPLQAEALSVNESHVTSRAMQDPVFQLPTAGRTPNGEVGAMDLTQLQKEKQDLEQQLLEKNKTIKQMQQRMLELRKTLQKELKIRPDNELFEVREKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC33154/golgin-97
Common Name GOLGA1
Gene Symbol GOLGA1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.