Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GOLGA5 Full-length ORF (NP_005104.2, 1 a.a. - 731 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946224 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-224 25 μg
89-946-223 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-224 Supplier Abnova™ Supplier No. H00009950P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins, whose members localize to the Golgi. This protein is a coiled-coil membrane protein that has been postulated to play a role in vesicle tethering and docking. Translocations involving this gene and the ret proto-oncogene have been found in tumor tissues; the chimeric sequences have been designated RET-II and PTC5. [provided by RefSeq]

Sequence: MSWFVDLAGKAEDLLNRVDQGAATALSRKDNASNIYSKNTDYTELHQQNTDLIYQTGPKSTYISSAADNIRNQKATILAGTANVKVGSRTPVEASHPVENASVPRPSSHFVRRKKSEPDDELLFDFLNSSQKEPTGRVEIRKEKGKTPVFQSSQTSSVSSVNPSVTTIKTIEENSFGSQTHEAASNSDSSHEGQEESSKENVSSNAACPDHTPTPNDDGKSHELSNLRLENQLLRNEVQSLNQEMASLLQRSKETQEELNKARARVEKWNADHSKSDRMTRGLRAQVDDLTEAVAAKDSQLAVLKVRLQEADQLLSTRTEALEALQSEKSRIMQDQSEGNSLQNQALQTFQERLHEADATLKREQESYKQMQSEFAARLNKVEMERQNLAEAITLAERKYSDEKKRVDELQQQVKLYKLNLESSKQELIDYKQKATRILQSKEKLINSLKEGSGFEGLDSSTASSMELEELRHEKEMQREEIQKLMGQIHQLRSELQDMEAQQVNEAESAREQLQDLHDQIAGQKASKQELETELERLKQEFHYIEEDLYRTKNTLQSRIKDRDEEIQKLRNQLTNKTLSNSSQSELENRLHQLTETLIQKQTMLESLSTEKNSLVFQLERLEQQMNSASGSSSNGSSINMSGIDNGEGTRLRNVPVLFNDTETNLAGMYGKVRKAASSIDQFSIRLGIFLRRYPIARVFVIIYMALLHLWVMIVLLTYTPEMHHDQPYGK

Specifications

Accession Number NP_005104.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9950
Molecular Weight (g/mol) 109.4kDa
Name GOLGA5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSWFVDLAGKAEDLLNRVDQGAATALSRKDNASNIYSKNTDYTELHQQNTDLIYQTGPKSTYISSAADNIRNQKATILAGTANVKVGSRTPVEASHPVENASVPRPSSHFVRRKKSEPDDELLFDFLNSSQKEPTGRVEIRKEKGKTPVFQSSQTSSVSSVNPSVTTIKTIEENSFGSQTHEAASNSDSSHEGQEESSKENVSSNAACPDHTPTPNDDGKSHELSNLRLENQLLRNEVQSLNQEMASLLQRSKETQEELNKARARVEKWNADHSKSDRMTRGLRAQVDDLTEAVAAKDSQLAVLKVRLQEADQLLSTRTEALEALQSEKSRIMQDQSEGNSLQNQALQTFQERLHEADATLKREQESYKQMQSEFAARLNKVEMERQNLAEAITLAERKYSDEKKRVDELQQQVKLYKLNLESSKQELIDYKQKATRILQSKEKLINSLKEGSGFEGLDSSTASSMELEELRHEKEMQREEIQKLMGQIHQLRSELQDMEAQQVNEAESAREQLQDLHDQIAGQKASKQELETELERLKQEFHYIEEDLYRTKNTLQSRIKDRDEEIQKLRNQLTNKTLSNSSQSELENRLHQLTETLIQKQTMLESLSTEKNSLVFQLERLEQQMNSASGSSSNGSSINMSGIDNGEGTRLRNVPVLFNDTETNLAGMYGKVRKAASSIDQFSIRLGIFLRRYPIARVFVIIYMALLHLWVMIVLLTYTPEMHHDQPYGK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GOLIM5/RFG5/ret-II
Common Name GOLGA5
Gene Symbol GOLGA5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.