Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GOLM1/GP-73 ELISA Kit

Catalog No. EH227RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH227RB 96 Tests
EH227RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH227RB Supplier Invitrogen™ Supplier No. EH227RB
Add to Cart
Add to Cart

ELISA

Human GOLM1/GP-73 quantitates human GOLM1/GP-73 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GOLM1/GP-73.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326
Assay Range 0.5 to 140 ng/mL
Assay Sensitivity 0.5 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 51280
Gene Symbol GOLM1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GOLPH2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.