Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GP1BA (NP_000164, 17 a.a - 306 a.a) Partial Protein (single sulfated form) with C-terminal hexahistidine tag expressed in HEK293EBNA1 cells

Catalog No. 89946309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
45 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-309 45 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-309 Supplier Abnova™ Supplier No. P5899
Add to Cart
Add to Cart

Used for SDS-PAGE

Glycoprotein Ib (GP Ib) is a platelet surface membrane glycoprotein composed of a heterodimer, an alpha chain and a beta chain, that are linked by disulfide bonds. The Gp Ib functions as a receptor for von Willebrand factor (VWF). The complete receptor complex includes noncovalent association of the alpha and beta subunits with platelet glycoprotein IX and platelet glycoprotein V. The binding of the GP Ib-IX-V complex to VWF facilitates initial platelet adhesion to vascular subendothelium after vascular injury, and also initiates signaling events within the platelet that lead to enhanced platelet activation, thrombosis, and hemostasis. This gene encodes the alpha subunit. Several polymorphisms and mutations have been described in this gene, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease. [provided by RefSeq]

Specifications

Accession Number NP_000164
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 2811
Molecular Weight (g/mol) 33.5kDa
Name GP1BA N37O/N175O (Human) Recombinant Protein
Preparation Method Mammalian cell (HEK293EBNA1) expression system
Quality Control Testing NuPAGE Stained with Coomassie Blue
Quantity 45 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BSS/CD42B/CD42b-alpha/GP1B/MGC34595
Common Name GP1BA
Gene Symbol GP1BA
Cross Reactivity Human ;Human
Species Human
Protein Tag His
Expression System Mammalian cell (HEK293EBNA1) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.