Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human GP1BB (NP_000398) VersaClone cDNA
SDP

Catalog No. RDC1820 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1820 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1820 Supplier R&D Systems™ Supplier No. RDC1820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glycoprotein lb [Platelet] beta.

The Human GP1BB (NP_000398) VersaClone cDNA clone contains the complete ORF for GP1BB, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GP1BB insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

Specifications

Product Type cDNA
Gene hGP1BB
Accession Number NP_000398
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 634bp
Gene ID (Entrez) 2812
Gene Alias antigen CD42b-beta, BDPLT1, BS, CD42c, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-Beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Show More Show Less

For Research Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.