Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human gp96/HSP90B1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB7606SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7606SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7606SP Supplier R&D Systems Supplier No. MAB7606SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

gp96/HSP90B1/GRP94 Monoclonal specifically detects gp96/HSP90B1/GRP94 in Hamster∣Human∣Mouse∣Rat∣Virus samples. It is validated for Gene Knock-Out∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Simple Western∣Western Blot.

Specifications

Antigen gp96/HSP90B1/GRP94
Applications Gene Knock-Out, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Simple Western, Western Blot
Classification Monoclonal
Clone 816803
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P14625
Gene Alias 94 kDa glucose-regulated protein, ECGP, Endoplasmin, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, Grp94, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, HSP90B1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96)
Gene Symbols HSP90B1
Host Species Mouse
Immunogen E. coli-derived recombinant human gp96/HSP90B1 Arg503-Arg660 Accession # P14625
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7184
Test Specificity Detects human gp96/HSP90B1 in direct ELISAs and Western blots. Detects mouse and rat gp96/HSP90B1 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Hamster, Human, Mouse, Rat, Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.