Learn More
ACROBiosystems ACROBiosystems Human GPA33 / A33 Protein, His Tag
Supplier:  ACROBiosystems GP3H52241MG
Encompass_Preferred
Recombinant Protein;1MG;Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GPA33, His Tag (Cat. No. GP3-H5224) at 5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPA33 Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-210-6652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.