Learn More
Invitrogen™ Human GPD2 Recombinant Protein
Description
This product contains purified recombinant human mitochondiral Glycerol-3-PO4 Dehydrogenase protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P43304 |
| Concentration | 0.095 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 1 mM EDTA, 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 2820 |
| Name | Human GPD2 |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.