Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GPM6B Full-length ORF (NP_001001996.1, 1 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-370 10 ug
89-946-371 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-370 Supplier Abnova™ Supplier No. H00002824P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a membrane glycoprotein that belongs to the proteolipid protein family. Proteolipid protein family members are expressed in most brain regions and are thought to be involved in cellular housekeeping functions, such as membrane trafficking and cell-to-cell communication. [provided by RefSeq]

Sequence: MKPAMETAAEENTEQSQERKVNSRAEMEIGRYHWMYPGSKNHQYHPVPTLGDRASPLSSPGCFECCIKCLGGVPYASLVATILCFSGVALFCGCGHVALAGTVAILEQHFSTNASDHALLSEVIQLMQYVIYGIASFFFLYGIILLAEGFYTTSAVKELHGEFKTTACGRCISGMFVFLTYVLGVAWLGVFGFSAVPVFMFYNIWSTCEVIKSPQTNGTTGVEQICVDIRQYGIIPWNAFPGKICGSALENICNTNEFYMSYHLFIVACAGAGATVIALLIYMMATTYNYAVLKFKSREDCCTKF

Specifications

Accession Number NP_001001996.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2824
Molecular Weight (g/mol) 59.9kDa
Name GPM6B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MKPAMETAAEENTEQSQERKVNSRAEMEIGRYHWMYPGSKNHQYHPVPTLGDRASPLSSPGCFECCIKCLGGVPYASLVATILCFSGVALFCGCGHVALAGTVAILEQHFSTNASDHALLSEVIQLMQYVIYGIASFFFLYGIILLAEGFYTTSAVKELHGEFKTTACGRCISGMFVFLTYVLGVAWLGVFGFSAVPVFMFYNIWSTCEVIKSPQTNGTTGVEQICVDIRQYGIIPWNAFPGKICGSALENICNTNEFYMSYHLFIVACAGAGATVIALLIYMMATTYNYAVLKFKSREDCCTKF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias M6B/MGC17150/MGC54284
Common Name GPM6B
Gene Symbol GPM6B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.