Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Gpr61 Full-Length Orf (Np_114142.3) Recombinant Protein Without Tag.

Catalog No. 89032157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-157 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-157 Supplier Abnova™ Supplier No. H00083873G01
Add to Cart
Add to Cart

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Good Folding

Immunogen Sequence: MESSPIPQSSGNSSTLGRVPQTPGPSTASGVPEVGLRDVASESVALFFMLLLDLTAVAGNAAVMAVIAKTPALRKFVFVFHLCLVDLLAALTLMPLAMLSSSALFDHALFGEVACRLYLFLSVCFVSLAILSVSAINVERYYYVVHPMRYEVRMTLGLVASVLVGVWVKALAMASVPVLGRVSWEEGAPSVPPGCSLQWSHSA

Specifications

Accession Number NP_114142.3
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 83873
Name GPR61 (Human) Recombinant Protein
Quantity 2 μg
Immunogen MESSPIPQSSGNSSTLGRVPQTPGPSTASGVPEVGLRDVASESVALFFMLLLDLTAVAGNAAVMAVIAKTPALRKFVFVFHLCLVDLLAALTLMPLAMLSSSALFDHALFGEVACRLYLFLSVCFVSLAILSVSAINVERYYYVVHPMRYEVRMTLGLVASVLVGVWVKALAMASVPVLGRVSWEEGAPSVPPGCSLQWSHSAYCQLFVVVFAVLYFLLPLLLILVVYCSMFRVARVAAMQHGPLPTWMETPRQRSESLSSRSTMVTSSGAPQTTPHRTFGGGKAAVVLLAVGGQFLLCWLPYFSFHLYVALSAQPISTGQVESVVTWIGYFCFTSNPFFYGCLNRQIRGELSKQFVCFFKPAPEEELRLPSREGSIEENFLQFLQGTGCPSESWVSRPLPSPKQEPPAVDFRIPGQIAEETSEFLEQQLTSDIIMSDSYLRPAASPRLES
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BALGR/GPCR3
Common Name GPR61
Gene Symbol GPR61
Species Wheat Germ (in vitro)
Protein Tag None
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.