Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GPSM2 Partial ORF (NP_037428, 381 a.a. - 480 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946501 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-501 25 μg
89-946-500 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-501 Supplier Abnova™ Supplier No. H00029899Q01L

Used for AP, Array, ELISA, WB-Re

Heterotrimeric G proteins transduce extracellular signals received by cell surface receptors into integrated cellular responses. GPSM2 belongs to a group of proteins that modulate activation of G proteins (Blumer et al., 2002 [PubMed 11832491]).[supplied by OMIM]

Sequence: ENTEIDSSLNGVRPKLGRRHSMENMELMKLTPEKVQNWNSEILAKQKPLIAKPSAKLLFVNRLKGKKYKTNSSTKVLQDASNSIDHRIPNSQRKISADTI

Specifications

Accession Number NP_037428
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29899
Molecular Weight (g/mol) 36.74kDa
Name GPSM2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ENTEIDSSLNGVRPKLGRRHSMENMELMKLTPEKVQNWNSEILAKQKPLIAKPSAKLLFVNRLKGKKYKTNSSTKVLQDASNSIDHRIPNSQRKISADTI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LGN/Pins
Common Name GPSM2
Gene Symbol GPSM2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.