Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Gremlin-1 Recombinant Protein, PeproTech®

Catalog No. 1204201M
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-042-01M 2 x 500 μg
12042100UG 100 μg
120-42-10UG 10 μg
120-42-1MG 2 x 500 μg
12042250UG 250 μg
12042500UG 500 μg
120-42-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 12-042-01M Supplier Gibco™ Supplier No. 1204201M
Add to Cart
Add to Cart

Recombinant Protein

120-42-1MG will be provided as 2 x 500 μg (120-42-500UG). Recombinant Human Gremlin-1 is a 18.3 kDa protein containing 160 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60565
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 26585
Molecular Weight (g/mol) 18.3 kDa
Name Human Gremlin-1
Purification Method Purified
Quantity 2 x 500 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C15DUPq; Cell proliferation-inducing gene 2 protein; CKTSF1B1; colorectal adenoma and carcinoma 1; CRAC1; CRCS4; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; DAND2; Down-regulated in Mos-transformed cells protein; down-regulated in v-mos-transformed cells; Drm; DUP15q; Grem; GREM1; gremlin; gremlin 1; gremlin 1 homolog, cysteine knot superfamily; gremlin 1, cysteine knot superfamily, homolog; gremlin 1, DAN family BMP antagonist; gremlin 1-like protein; gremlin-1; HMPS; HMPS1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2; ld; MPSH; PIG2
Common Name GREM1
Gene Symbol Grem1
Biological Activity Determined by its ability to inhibit BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The ED50 for this effect is 0.07-0.11 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence KKKGSQGAIP PPDKAQHNDS EQTQSPQQPG SRNRGRGQGR GTAMPGEEVL ESSQEALHVT ERKYLKRDWC KTQPLKQTIH EEGCNSRTII NRFCYGQCNS FYIPRHIRKE EGSFQSCSFC KPKKFTTMMV TLNCPELQPP TKKKRVTRVK QCRCISIDLD
Content And Storage -20°C
Activity Determined by its ability to inhibit BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The ED50 for this effect is 0.07-0.11 ug/ml.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.