Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRID1 Partial ORF (NP_060021, 349 a.a. - 441 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946567 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-567 25 ug
89-946-566 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-567 Supplier Abnova™ Supplier No. H00002894Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a subunit of glutamate receptor channels. These channels mediate most of the fast excitatory synaptic transmission in the central nervous system and play key roles in synaptic plasticity

Sequence: LNCIRKSTKPWNGGRSMLDTIKKGHITGLTGVMEFREDSSNPYVQFEILGTTYSETFGKDMRKLATWDSEKGLNGSLQERPMGSRLQGLTLKV

Specifications

Accession Number NP_060021
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2894
Molecular Weight (g/mol) 35.97kDa
Name GRID1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen LNCIRKSTKPWNGGRSMLDTIKKGHITGLTGVMEFREDSSNPYVQFEILGTTYSETFGKDMRKLATWDSEKGLNGSLQERPMGSRLQGLTLKV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA1220
Common Name GRID1
Gene Symbol GRID1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.