Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human GRO-gamma (CXCL3) Recombinant Protein, PeproTech®

Catalog No. 30040100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30040100UG 100 μg
300-40-10UG 10 μg
300-40-1MG 1 mg
30040250UG 250 μg
300-40-2UG 2 μg
30040500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30040100UG Supplier Gibco™ Supplier No. 30040100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GRO-gamma is a 7.9 kDa protein consisting of 73 amino acids including the 'ELR' motif common to the CXC chemokine family that bind to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19876
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2921
Molecular Weight (g/mol) 7.9 kDa
Name Human GRO-gamma (CXCL3)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2 b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2 b; MIP2-beta; SCYB3
Common Name CXCL3
Gene Symbol Cxcl3
Biological Activity Determined by its ability to chemoattract 293 transfected CXCR2 cells using a concentration range of 10-100 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.