Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Growth Hormone Recombinant Protein

Catalog No. PIRP10928
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP10928 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP10928 Supplier Invitrogen™ Supplier No. RP10928
Add to Cart
Add to Cart

Recombinant Protein

This is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Store lyophilized product desiccated at -20° C. Upon reconstitution, store at 4° C for 2-7 days or aliquot and store at -20° C. For long term storage add either HSA or BSA to 0.1%. Avoid repeated freeze/thaw cycles. Reconstitute using distilled water to a concentration not less than 100 μg/mL.

Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01241
Concentration 100 μg/mL
For Use With (Application) Control
Formulation protein with no preservative
Gene ID (Entrez) 2688
Molecular Weight (g/mol) 22.260 kDa
Name Human Growth Hormone
Quantity 1000 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; RNGHGP; Somatotropin
Common Name Growth Hormone
Gene Symbol GH1
Biological Activity Fully biologically active as compared to WHO reference standard at 3 U/mg
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human Growth Hormone recombinant protein contains 192 amino acids
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.