Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRSF1 Partial ORF (NP_002083, 87 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946654 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-654 10 ug
89-946-655 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-654 Supplier Abnova™ Supplier No. H00002926Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a cellular protein that binds RNAs containing the G-rich element. The protein is localized in the cytoplasm, and has been shown to stimulate translation of viral mRNAs in vitro. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: EEEVDDVFLIRAQGLPWSCTMEDVLNFFSDCRIRNGENGIHFLLNRDGKRRGDALIEMESEQDVQKALEKHRMYMGQRYVEVYEINNEDVDALMKSLQ

Specifications

Accession Number NP_002083
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2926
Molecular Weight (g/mol) 36.52kDa
Name GRSF1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen EEEVDDVFLIRAQGLPWSCTMEDVLNFFSDCRIRNGENGIHFLLNRDGKRRGDALIEMESEQDVQKALEKHRMYMGQRYVEVYEINNEDVDALMKSLQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ13125
Common Name GRSF1
Gene Symbol GRSF1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.