Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GSK3B (P49841) Full-length Recombinant Protein with His tag

Catalog No. 89946686 Shop All Abnova Corporation Products
Change view
Click to view available options
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-946-686 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-686 Supplier Abnova™ Supplier No. P4183

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE

The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene

Specifications

Accession Number P49841
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 2932
Molecular Weight (g/mol) 53.0-56.0kDa
Name GSK3B (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 100 ug
Storage Requirements Store at -80°C on dry atmosphere. After reconstitution with PBS containing 15% glycerol, store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GSK3B
Gene Symbol GSK3B
Biological Activity Activity of GSK-3beta was determined by an in vitro kinase assay using 100ng Tau as substrate and 1μg of the recombinant GSK-3beta in a 20μL reaction containing 25mM Tris-HCl, pH 7.5, 5mM beta-glycerol phosphate, 12mM MgCl2, 2mM DTT, 0.1
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.