Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Guanylyl Cyclase C/GUCY2C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7199046
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2157SP 25 μg
MAB2157100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2157SP Supplier R&D Systems Supplier No. MAB2157SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GUCY2C Monoclonal specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen GUCY2C
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 2543C
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P25092
Gene Alias DIAR6, EC 4.6.1, GCC, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylate Cyclase C, Guanylyl cyclase C, GUC2C, GUC2CEC 4.6.1.2, GUCY2C, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, MUCIL, STA Receptor, StAR, STARSTA receptor
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2984
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.