Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HAAO Full-length ORF (AAH29510.1, 1 a.a. - 286 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-040 10 μg
89-947-041 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-040 Supplier Abnova™ Supplier No. H00023498P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

3-Hydroxyanthranilate 3,4-dioxygenase is a monomeric cytosolic protein belonging to the family of intramolecular dioxygenases containing nonheme ferrous iron. It is widely distributed in peripheral organs, such as liver and kidney, and is also present in low amounts in the central nervous system. HAAO catalyzes the synthesis of quinolinic acid (QUIN) from 3-hydroxyanthranilic acid. QUIN is an excitotoxin whose toxicity is mediated by its ability to activate glutamate N-methyl-D-aspartate receptors. Increased cerebral levels of QUIN may participate in the pathogenesis of neurologic and inflammatory disorders. HAAO has been suggested to play a role in disorders associated with altered tissue levels of QUIN. [provided by RefSeq]

Sequence: MERRLGVRAWVKENRGSFQPPVCNKLMHQEQLKVMFIGGPNTRKDYHIEEGEEVFYQLEGDMVLRVLEQGKHRDVVIRQGEIFLLPARVPHSPQRFANTVGLVVERRRLETELDGLRYYVGDTMDVLFEKWFYCKDLGTQLAPIIQEFFSSEQYRTGKPIPDQLLKEPPFPLSTRSIMEPMSLDAWLDSHHRELQAGTPLSLFGDTYETQVIAYGQGSSEGLRQNVDVWLWQLEGSSVVTMGGRRLSLAPDDSLLVLAGTSYAWERTQGSVALSVTQDPACKKPLG

Specifications

Accession Number AAH29510.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23498
Molecular Weight (g/mol) 59kDa
Name HAAO (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MERRLGVRAWVKENRGSFQPPVCNKLMHQEQLKVMFIGGPNTRKDYHIEEGEEVFYQLEGDMVLRVLEQGKHRDVVIRQGEIFLLPARVPHSPQRFANTVGLVVERRRLETELDGLRYYVGDTMDVLFEKWFYCKDLGTQLAPIIQEFFSSEQYRTGKPIPDQLLKEPPFPLSTRSIMEPMSLDAWLDSHHRELQAGTPLSLFGDTYETQVIAYGQGSSEGLRQNVDVWLWQLEGSSVVTMGGRRLSLAPDDSLLVLAGTSYAWERTQGSVALSVTQDPACKKPLG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 3-HAO/HAO
Common Name HAAO
Gene Symbol HAAO
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.