Learn More
Sino Biological Human HAVCR1 PAb 100UG 11051-RP04-B-100
Supplier:  Sino Biological 11051RP04B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HAVCR1 / TIM1 / KIM1 (rh HAVCR1 / TIM1 / KIM1; Catalog#11051-H07H; AAC39862.1; Ser 21-Gly 290). HAVCR1 / TIM1 / KIM1 specific IgG was purified by Human HAVCR1 / TIM1 / KIM1 affinity chromatography and then biotinylated.
Catalog No. 50-241-2092
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.