Learn More
Description
Specifications
Specifications
| Assay Range | 7.81 - 500pg/mL |
| Assay Sensitivity | 1.74pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested |
| Sample Type | Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human HB-EGF |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Serum (100uL),EDTA Plasma (100uL),Platelet-poor EDTA Plasma (100uL),Heparin Plasma (100uL),Platelet-poor Heparin Plasma (100uL),Saliva (50uL),Urine (100uL),Human Milk (100uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
