Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human hCG ELISA Kit (Colorimetric)
SDP

Catalog No. NB600381KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600381KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600381KT Supplier Novus Biologicals™ Supplier No. NBP2600381KIT
Only null left
Add to Cart
Add to Cart

Assay has high sensitivity and specificity for detection of human HCG

The Human Chorionic Gonadotropin beta Chain (hCG beta) ELISA Kit (Colorimetric) measures human Chorionic Gonadotropin beta Chain (hCG beta) in serum, plasma, urine, cell culture supernates.

Specifications

Assay Range 8 to 240mIU/mL
Assay Sensitivity 2 mIU/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Specificity For detection of human HCG
Target Species Human
For Use With (Application) ELISA
Kit Contents Adhesive Strip (For 96 wells), Assay plate, HRP-conjugate, Wash Buffer (20 x concentrate), Substrate A, Substrate B, Stop Solution, Standard, Sample Diluent (for urine only, 10 x concentrate), Instruction manual
Quantity 1 Kit
Research Discipline Cancer
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target hCG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.