Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HDAC9 Full-length ORF (NP_848512.1, 1 a.a. - 1069 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-209 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-947-209 Supplier Abnova™ Supplier No. H00009734P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to the Xenopus and mouse MITR genes. The MITR protein lacks the histone deacetylase catalytic domain. It represses MEF2 activity through recruitment of multicomponent corepressor complexes that include CtBP and HDACs. This encoded protein may play a role in hematopoiesis. Multiple alternatively spliced transcripts have been described for this gene but the full-length nature of some of them has not been determined. [provided by RefSeq]

Sequence: MHSMISSVDVKSEVPVGLEPISPLDLRTDLRMMMPVVDPVVREKQLQQELLLIQQQQQIQKQLLIAEFQKQHENLTRQHQAQLQEHIKLQQELLAIKQQQELLEKEQKLEQQRQEQEVERHRREQQLPPLRGKDRGRERAVASTEVKQKLQEFLLSKSATKDTPTNGKNHSVSRHPKLWYTAAHHTSLDQSSPPLSGTSPSYKYTLPGAQDAKDDFPLRKTASEPNLKVRSRLKQKVAERRSSPLLRRKDGNVVTSFKKRMFEVTESSVSSSSPGSGPSSPNNGPTGSVTENETSVLPPTPHAEQMVSQQRILIHEDSMNLLSLYTSPSLPNITLGLPAVPSQLNASNSLKEKQKCETQTLRQGVPLPGQYGGSIPASSSHPHVTLEGKPPNSSHQALLQHLLLKEQMRQQKLLVAGGVPLHPQSPLATKERISPGIRGTHKLPRHRPLNRTQSAPLPQSTLAQLVIQQQHQQFLEKQKQYQQQIHMNKLLSKSIEQLKQPGSHLEEAEEELQGDQAMQEDRAPSSGNSTRSDSSACVDDTLGQVGAVKVKEEPVDSDEDAQIQEMESGEQAAFMQQPFLEPTHTRALSVRQAPLAAVGMDGLEKHRLVSRTHSSPAASVLPHPAMDRPLQPGSATGIAYDPLMLKHQCVCGNSTTHPEHAGRIQSIWSRLQETGLLNKCERIQGRKASLEEIQLVHSEHHSLLYGTNPLDGQKLDPRILLGDDSQKFFSSLPCGGLGVDSDTIWNELHSSGAARMAVGCVIELASKVASGELKNGFAVVRPPGHHAEESTAMGFCFFNSVAITAKYLRDQLNISKILIVDLDVHHGNGTQQAFYADPSILYISLHRYDEGNFFPGSGAPNEVGTGLGEGYNINIAWTGGLDPPMGDVEYLEAFRTIVKPVAKEFDPDMVLVSAGFDALEGHTPPLGGYKVTAKCFGHLTKQLMTLADGRVVLALEGGHDLTAICDASEACVNALLGNELEPLAEDILHQSPNMNAVISLQKIIEIQSKYWKSVRMVAVPRGCALAGAQLQEETETVSALASLTVDVEQPFAQEDSRTAGEPMEEEPAL

Specifications

Accession Number NP_848512.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9734
Molecular Weight (g/mol) 144kDa
Name HDAC9 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MHSMISSVDVKSEVPVGLEPISPLDLRTDLRMMMPVVDPVVREKQLQQELLLIQQQQQIQKQLLIAEFQKQHENLTRQHQAQLQEHIKLQQELLAIKQQQELLEKEQKLEQQRQEQEVERHRREQQLPPLRGKDRGRERAVASTEVKQKLQEFLLSKSATKDTPTNGKNHSVSRHPKLWYTAAHHTSLDQSSPPLSGTSPSYKYTLPGAQDAKDDFPLRKTASEPNLKVRSRLKQKVAERRSSPLLRRKDGNVVTSFKKRMFEVTESSVSSSSPGSGPSSPNNGPTGSVTENETSVLPPTPHAEQMVSQQRILIHEDSMNLLSLYTSPSLPNITLGLPAVPSQLNASNSLKEKQKCETQTLRQGVPLPGQYGGSIPASSSHPHVTLEGKPPNSSHQALLQHLLLKEQMRQQKLLVAGGVPLHPQSPLATKERISPGIRGTHKLPRHRPLNRTQSAPLPQSTLAQLVIQQQHQQFLEKQKQYQQQIHMNKLLSKSIEQLKQPGSHLEEAEEELQGDQAMQEDRAPSSGNSTRSDSSACVDDTLGQVGAVKVKEEPVDSDEDAQIQEMESGEQAAFMQQPFLEPTHTRALSVRQAPLAAVGMDGLEKHRLVSRTHSSPAASVLPHPAMDRPLQPGSATGIAYDPLMLKHQCVCGNSTTHPEHAGRIQSIWSRLQETGLLNKCERIQGRKASLEEIQLVHSEHHSLLYGTNPLDGQKLDPRILLGDDSQKFFSSLPCGGLGVDSDTIWNELHSSGAARMAVGCVIELASKVASGELKNGFAVVRPPGHHAEESTAMGFCFFNSVAITAKYLRDQLNISKILIVDLDVHHGNGTQQAFYADPSILYISLHRYDEGNFFPGSGAPNEVGTGLGEGYNINIAWTGGLDPPMGDVEYLEAFRTIVKPVAKEFDPDMVLVSAGFDALEGHTPPLGGYKVTAKCFGHLTKQLMTLADGRVVLALEGGHDLTAICDASEACVNALLGNELEPLAEDILHQSPNMNAVISLQKIIEIQSKYWKSVRMVAVPRGCALAGAQLQEETETVSALASLTVDVEQPFAQEDSRTAGEPMEEEPAL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp779K1053/HD7/HDAC/HDAC7/HDAC7B/HDAC9B/HDAC9FL/HDRP/KIAA0744/MITR
Common Name HDAC9
Gene Symbol HDAC9
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.