Learn More
Description
- Human heart atrium, left tissue lysate was prepared by homogenization using a proprietary technique
- The tissue was frozen in liquid nitrogen immediately after excision and then stored at -70°C
- The human heart atrium, left tissue total protein is provided in a buffer including HEPES (pH7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
- For quality control purposes, the heart atrium, left tissue pattern on SDS-PAGE gel is shown to be consistent for each lot by visualization with coomassie blue staining
- The heart atrium, left tissue is then Western analyzed by either GAPDH or β-actin antibody, and the expression level is consistent with each lot.
Specifications
Specifications
| Quantity | 0.1 mg |
| Tissue Condition | Diseased |
| Host Species | Human |
| Concentration | 2 mg/mL |
| Protein State | Denatured |
| Tissue | Heart, Left Atrium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
